Order Entry
Denmark
ContactUsLinkComponent
TRIS-buffered saline (TBS, 10X) pH 7,4
TRIS-buffered saline (TBS, 10X) pH 7,4
Certificates
TRIS-buffered saline (TBS, 10X) pH 7,4
Certificates

Specifications

  • MDL No.:
    MFCD00132476
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

Related Information

Frequently Bought Together