Neuraminidase (from Clostridium perfringens)
Catalog # J60711.LR
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 U/g
- CAS No.:9001-67-6
- Form:Lyophilized powder
- MDL No.:MFCD00131711
- Enzyme name:Neuraminidase
Specifications
About this item
Applications: A hydrolytic enzyme that removes sialic acid from mucoproteins