Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

C2 Ceramide 98%

10 mg
Catalog # J60534.MA
C2 Ceramide 98%
Catalog # J60534.MA
CAS Number:  3102-57-6
C2 Ceramide 98%
C2 Ceramide 98%

10 mg

CAS Number:  3102-57-6
Price
1.970,00 kr
Log in for availability
1 * 10 mg
Product Details & Documents

Purity: 98% CAS No.: 3102-57-6 Molecular Formula: C20H39NO3 Formula Weight: 341.53 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 77-78° Solubility: Soluble in ethanol, methanol, chloroform or DMSO at approximately 10mg/ml MDL No.: MFCD00153903

Applications: Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases

  • CAS Number:3102-57-6
  • Formula:C₂₀H₃₉NO₃
  • MDL Number:MFCD00153903
  • Molecular Weight:341.536 g/mol
  • Melting Pt:77…78 °C
Specifications

Specifications

  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.