Order Entry
Denmark
ContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use, precipitating
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use, precipitating
Catalog # J60461.AP
CAS Number:  
3,3',5,5'-Tetramethylbenzidine (TMB) solution, standard sensitivity ready-to-use, precipitating
Catalog # J60461.AP
CAS Number:  

Specifications

  • Pk:
    500 mL

Specifications

About this item

CAS Number:54827-17-7
 :
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748
 :240.35 g/mol
Storage Temperature:Refrigerator
EINECS:259-364-6
UN:2810
ADR:6.1,III
 :GHS06
 :GHS08