3,5-Bis(methoxycarbonyl)phenylboronic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:177735-55-6
:
Formula:C₁₀H₁₁BO₆
MDL Number:MFCD11053854
:238.00 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together