Order Entry
Denmark
ContactUsLinkComponent
5-(Trifluoromethyl)pyridine-3-carboxylic acid 97%
Irritant
5-(Trifluoromethyl)pyridine-3-carboxylic acid 97%
Catalog # H63906.03
CAS Number:  
Irritant
5-(Trifluoromethyl)pyridine-3-carboxylic acid 97%
Catalog # H63906.03
CAS Number:  

Specifications

  • Pk:
    1 g

Specifications

About this item

CAS Number:131747-40-5
 :
Formula:C₇H₄F₃NO₂
 :191.11 g/mol
Melting Pt:184…189 °C
Storage Temperature:Ambient
ADR:6.1,III
 :GHS07
MDL Number:MFCD07774128