


(S)-(-)-2,3-Epoxy-1-propanol ≥99%, ee 99% ee
Leverandør: Thermo Fisher Scientific
Certifikater
Om denne vare
CAS nummer:60456-23-7
:
Formel:C₃H₆O₂
MDL nummer:MFCD00074874
:74.08 g/mol
Kogende Pt:66…67 °C (19 mmHg)
Tæthed:1.116
Flash Pt:66 °C (151 °F)
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Relateret information
Frequently Bought Together