
2-Amino-3,3,3-trifluoropropionic acid ethylester hydrochloride 96%
Catalog # H33319.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.58 g/mol
Melting Pt:∼ 149 °C. (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:193140-71-5
Formula:C₅H₉ClF₃NO₂
MDL Number:MFCD14525479