Order Entry
Denmark
ContactUsLinkComponent
5-Bromo-6-chloronicotinic acid 97%
Irritant
5-Bromo-6-chloronicotinic acid 97%
Catalog # H32384.06
CAS Number:  
Irritant
5-Bromo-6-chloronicotinic acid 97%
Catalog # H32384.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

CAS Number:29241-62-1
 :
Formula:C₆H₃BrClNO₂
 :236.45 g/mol
Melting Pt:170…173 °C
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD01927098