Order Entry
Denmark
ContactUsLinkComponent
2-Chloro-4-(trifluoromethyl)nicotinic acid 97%
Irritant
2-Chloro-4-(trifluoromethyl)nicotinic acid 97%
Catalog # H27312.03
CAS Number:  
Irritant
2-Chloro-4-(trifluoromethyl)nicotinic acid 97%
Catalog # H27312.03
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:158…159 °C
 :225.56 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:590371-81-6
Formula:C₇H₃ClF₃NO₂
MDL Number:MFCD08458125