Tungsten, gauze woven from wire 150 mesh woven from 0.02mm (0.0008in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-33-7
:
Formula:W
MDL Number:MFCD00011461
:183.84 g/mol
Storage Temperature:Ambient
EINECS:231-143-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together