Order Entry
Denmark
ContactUsLinkComponent
 
 
 
 
n-Pentane, anhydrous ≥99.8%, over molecular sieves
  47115.AK
 :  
undefined
 
 
 
n-Pentane, anhydrous ≥99.8%, over molecular sieves
  47115.AK
 :  

 

  • Pk:
    250 mL
  • Seq:
    2590

 

 

 :
 :72.15 g/mol
 :14,7116
 :36 °C
 :–130 °C
 :0.626
 :–49 °C (–57 °F)
 :Ambient
 :203-692-4
 :1265
 :3,II
 :Packaged under argon in resealable ChemSeal™ bottles.
 :GHS02
 :GHS08
 :GHS07
 :109-66-0
 :H₃C(CH₂)₃CH₃
 :MFCD00009498