Order Entry
Denmark
ContactUsLinkComponent
Nickel 61, 61 Ni, plasma standard solution, 61 Ni 10 µg/ml, Specpure®
Nickel 61, 61 Ni, plasma standard solution, 61 Ni 10 µg/ml, Specpure®
Catalog # 45282.AD
CAS Number:  
Nickel 61, 61 Ni, plasma standard solution, 61 Ni 10 µg/ml, Specpure®
Catalog # 45282.AD
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    50 mL
  • Description:
    Nickel 61 isotope
  • MDL No.:
    MFCD00144901
  • Element:
    Nickel
  • Symbol:
    Ni

Specifications

About this item

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00144901