Order Entry
Denmark
ContactUsLinkComponent
Lead(II) metaborate monohydrate
Health
Irritant
Lead(II) metaborate monohydrate
Catalog # 39339.22
CAS Number:  
Health
Irritant
Lead(II) metaborate monohydrate
Catalog # 39339.22
CAS Number:  

Specifications

  • Pk:
    100 g

Specifications

About this item

 :
Melting Pt:160 °C (-H₂O)
Density:5.598
EINECS:238-762-3
ADR:6.1,III
CAS Number:10214-39-8