Order Entry
Denmark
ContactUsLinkComponent
Palladium 0.5% on alumina, pellets 3.18 mm (0.125 in), reduced
Palladium 0.5% on alumina, pellets 3.18 mm (0.125 in), reduced
Catalog # 38786.14
CAS Number:  
Palladium 0.5% on alumina, pellets 3.18 mm (0.125 in), reduced
Catalog # 38786.14
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :106.420 g/mol
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167