Aluminum phosphate ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7784-30-7
:
Formula:AlPO₄
MDL Number:MFCD00003430
:121.951 g/mol
Merck Index:14,358
Melting Pt:>1500 °C
Density:2.566
Storage Temperature:Ambient
EINECS:232-056-9
UN:3260
ADR:8,III
:GHS05
Frequently Bought Together