Anti-CD40 Goat Polyclonal Antibody
EVBIEB11699
- Antibody type:Primary
- Antigen name:CD40 molecule, TNF receptor superfamily member 5
- Antigen symbol:CD40
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:affinity purified
- Form:liquid
- Gene ID:958
- Antigen synonyms:p50|Bp50|TNFRSF5|CDW40
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sequence:C-PQEINFPDDLPGSN
- Storage temperature:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Concentration:100µg/200µl
- Shipping temperature:8 °C
- Purification:Affinity purified
- Size:100µg
- Pack type:plastic vial
- Pk:0,1 mg
Western Blot: Approx 48kDa band observed in lysates of cell line Daudi (calculated MW of 30.6kDa according to NP_001241.1). The observed molecular weight corresponds to the glycosylated form. Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human