Order Entry
Denmark
ContactUsLinkComponent
Anti-GOLT1B Rabbit Polyclonal Antibody
Anti-GOLT1B Rabbit Polyclonal Antibody
Catalog # ABGEAP13094B
Supplier:  Abgent
Anti-GOLT1B Rabbit Polyclonal Antibody
Catalog # ABGEAP13094B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    golgi transport 1B
  • Antigen symbol:
    GOLT1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    51026
  • Antigen synonyms:
    YMR292W|CGI-141|GCT2|GOT1|GOT1B
  • Amino acid number:
    108 to 136
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    15.426 kDa
  • Sequence:
    GIRSFVDKVGESNNC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.61
  • Shipping temperature:
    4 °C
  • Immunogen:
    This GOT1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 108-136 amino acids from the C-terminal region of human GOT1B.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: GOLT1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse