Order Entry
Denmark
Orders LinkContactUsLinkComponent
Storage vials
Storage vials
Supplier:   SAVILLEX
Storage vials
Supplier:  SAVILLEX
Certificates
Storage vials
4 Options Available Below
Product Details & Documents
PFA vials with round or conical interior.
  • Ideal for storage and digestion in geochemistry
  • Flat base for maximum heating efficiency
  • Square exterior vials with hex closures have higher pressure rating
Ordering information: Closures can be ordered separately. Modifications may be available on request.
Product Family Options
Product Information
  • Capacity
    Ø×H
    Cap included
    Cap Size
    Availability
    Price
  • SAVI200-003-20
    Standard vial with round interior and square exterior
    Standard vial with round interior and square exterior
    3,00 ml
    17,3×26,4 mm
    No
    23 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 167,00 kr
    167,00 kr
    1 * 1 Items
       167,00 kr
  • SAVI200-007-20
    Standard vial with round interior and round exterior
    Standard vial with round interior and round exterior
    7,00 ml
    22,1×37,1 mm
    No
    24 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 201,00 kr
    201,00 kr
    1 * 1 Items
       201,00 kr
  • SAVI200-015-20
    Standard vial with round interior and round exterior
    Standard vial with round interior and round exterior
    15,00 ml
    30,5×41,4 mm
    No
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 351,00 kr
    351,00 kr
    1 * 1 Items
       351,00 kr
  • SAVI200-030-12
    Standard vial with flat interior and round exterior
    Standard vial with flat interior and round exterior
    30,00 ml
    30,5×69,9 mm
    No
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 538,00 kr
    538,00 kr
    1 * 1 Items
       538,00 kr
Closures
Product Information
  • Catalog #
    Type
    For
    Cap Size
    Availability
    Price
  • PFA, plain threaded
    Micro columns, tubes and vials
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 201,00 kr
    201,00 kr
    1 * 1 Items
       201,00 kr
Product Information
  • Catalog #
    Type
    For
    Cap Size
    Availability
    Price
  • PFA, plain hex threaded
    Micro columns, tubes and vials
    23 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 151,00 kr
    151,00 kr
    1 * 1 Items
       151,00 kr
Product Information
  • Catalog #
    Type
    For
    Cap Size
    Availability
    Price
  • PFA, plain threaded
    Micro columns, tubes, standard and autosampler vials
    24 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 151,00 kr
    151,00 kr
    1 * 1 Items
       151,00 kr