Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Sodium thiosulphate solution 2.12 N

100 mL
Catalog # HACH2686832
Supplier:   Hach
Sodium thiosulphate solution 2.12 N
Catalog # HACH2686832
Supplier:  Hach
CAS Number:  7772-98-7
Sodium thiosulphate solution 2.12 N
Sodium thiosulphate solution 2.12 N

100 mL

CAS Number:  7772-98-7
Price
518,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

For determination of high range hypochlorite (bleach) concentrations by drop-count titration.

Synonyms:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS Number:7772-98-7
  • Formula:Na₂S₂O₃
  • MDL Number:MFCD00003499
  • Molecular Weight:158.11 g/mol
Specifications

Specifications

  • Pk:
    100 mL

Specifications