Order Entry
Denmark
Orders LinkContactUsLinkComponent
Erlenmeyer flasks, narrow neck
Erlenmeyer flasks, narrow neck
Leverandør:   DWK Life Sciences
Erlenmeyer flasks, narrow neck
Leverandør:  DWK Life Sciences
Certificates
Erlenmeyer flasks, narrow neck
2 Options Available Below
Product Details & Documents
Borosilicate glass 3,3, KIMAX®.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With graduation.
Certifications: DIN ISO 1773
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Neck I-Ø
    Tilgængelighed
    Price
  • 25 ml
    140 mm
    85 mm
    34 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 346,00 kr
    346,00 kr
    1 * 10 Items
       346,00 kr
  • 250 ml
    140 mm
    85 mm
    34 mm
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 300,00 kr
    300,00 kr
    1 * 10 Items
       300,00 kr