Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cadmium standard solution

Cadmium standard solution

Cadmium standard solution (NIST), 100 mg/l as Cd, 100 mL
Catalog # HACH1402442
Supplier:   Hach
Cadmium standard solution
Catalog # HACH1402442
Supplier:  Hach
CAS Number:  
Cadmium standard solution
Cadmium standard solution

Cadmium standard solution (NIST), 100 mg/l as Cd, 100 mL

CAS Number:  
Price
310,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in cadmium determinations.

Specifications

Specifications

  • Concentration:
    100 mg/l as Cd
  • Pk:
    100 mL
  • Description:
    Cadmium standard solution (NIST)

Specifications