Order Entry
Denmark
ContactUsLinkComponent
 
Glial cell line-derived neurotrophic factor (GDNF) Rapid™ ELISA kit: mouse
 :  Biosensis
 
undefined
Glial cell line-derived neurotrophic factor (GDNF) Rapid™ ELISA kit: mouse
 :  Biosensis

 

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    GDNF
  • Cross reactivity:
    No cross-reactivity was observed for the following proteins tested at 25 ng/mL: brain-derived neurotrophic factor (rhBDNF), nerve growth factor (rhNGF), neurotrophin-3 (rhNT-3), rhNT-4/5, vascular endothelial growth factor (rhVEGF), recombinant human Neurturin, Artemin and Persephin.
  • Specificity:
    Mouse. The antibodies used in this kit detect human and rat GDNF.
  • Detection range:
    7,8 – 500 pg/ml
  • Sensitivity:
    Typical limit of detection (LOD) for rmGDNF is <10 pg/ml determined as blank value plus 3× standard deviation of blank OD (n=10).

 

Frequently Bought Together