Order Entry
Denmark
ContactUsLinkComponent
Multimodal chromatography columns, HiScreen™ Capto MMC ImpRes
Multimodal chromatography columns, HiScreen™ Capto MMC ImpRes
Catalog # 17-3716-20
Supplier:  Cytiva
undefined
Multimodal chromatography columns, HiScreen™ Capto MMC ImpRes
Catalog # 17-3716-20
Supplier:  Cytiva

Specifications

  • Particle Size:
    40 µm
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ Capto MMC ImpRes
  • pH range:
    3 - 12
  • Column hardware:
    PP
  • Bed dimensions:
    7,7×100 mm
  • Pk:
    1

Specifications

About this item

HiScreen Capto MMC ImpRes is a ready-to-use column pre-packed with BioProcess Capto MMC ImpRes weak cation exchange multimodal resin. The columns are an excellent choice for method optimization and parameter screening.

  • High yields achieved through the high-resolution beads and selectivity of the ligand
  • Enables use of a platform approach to MAb process development
  • Displays a broad pH/salt operational window
  • 10 cm bed height of HiScreen columns allows method optimisation and parameter screening
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

The multimodal functionality of Capto MMC ImpRes gives a different selectivity compared with traditional ion exchange columns and it binds proteins at high or low ionic strength.

Contaminants such as DNA, host cell proteins (HCP), leached protein A, and aggregates, are efficiently removed, making Capto MMC ImpRes an excellent choice for purification of MAbs and other biomolecules with high aggregate content.

Capto MMC ImpRes can typically be used in the initial polishing step in MAb purification processes, as well as for polishing of antibody fragments such as domain antibodies (DAbs).