Safety Funnel, for drums
Supplier: JUSTRITE MANUFACTURING
Certificates
About this item
Galvanised steel, 18-gauge, with electrostatically applied epoxy powder coat paint.
- Wide mouth opening, stay-opened lid
- Self-closing lin incase of fire
- Built-in flame arrester on most models to prevent fire intrusion
- Funnels for use with viscous liquids are fitted with brass tube
These safety drum funnels are mainly used to prevent spills while transferring flammable solvents, thinners, oils or paint wastes.
Approved by FM and compliance with EPA and OSHA regulations.
Frequently Bought Together