- Antibody type:Primary
- Antigen name:Microtubule-associated Protein 1 light chain 3 beta
- Antigen symbol:LC3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Gene ID:67443, 643246, 64862, 81631
- Antigen synonyms:MAP1A/1BLC3|autophagy-related ubiquitin-like modifier LC3 B|MAP1A/MAP1B light chain 3 B|MAP1 light chain 3-like protein 2|MAP1A/MAP1B LC3 B|ATG8F|LC3B|microtubule-associated proteins 1A/1B light chain 3B
- Storage buffer:PBS, 20% glycerol, 1% BSA, 0.025% Proclin 300, pH 7
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:0.43 mg/ml
- Immunogen:Synthetic peptide corresponding to a region within amino acids 1 and 125 of Human LC3B
- Purification:Antigen affinity chromatography
- Size:100 µL
- Pk:100 µl
This Antibody targets LC3B1+LC3B2 in WB applications and shows reactivity with Human samples. The immunogen is synthetic peptide corresponding to a region within amino acids 1 and 125 of Human LC3B. The product of this gene is a subunit of neuronal microtubule-associated MAP1A and MAP1B proteins, which are involved in microtubule assembly and important for neurogenesis. Studies on the rat homolog implicate a role for this gene in autophagy, a process that involves the bulk degradation of cytoplasmic component.
Western Blot: 1:1000-1:10,000, Immunofluorescence: 1:100-1:1000, Immunocytochemistry: 1:100-1:1000, Flow Cytometry: Assay-Dependent
Type: Primary
Antigen: LC3B1+LC3B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat