Order Entry
Denmark
Orders LinkContactUsLinkComponent
Anti-GPRIN2 Rabbit Polyclonal Antibody

Anti-GPRIN2 Rabbit Polyclonal Antibody

100 μg
Catalog # BOSSBS-8276R
Supplier:   Bioss
Anti-GPRIN2 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8276R
Supplier:  Bioss
Anti-GPRIN2 Rabbit Polyclonal Antibody

100 μg

Price
4.870,00 kr
Log in for availability
1 * 100 µl
Product Details & Documents

G protein-coupled receptors (GPCRs) represent a large superfamily of cell-surface receptors that are involved in a multitude of physiological processes such as perception of sensory information, modulation of synaptic transmission, hormone release/actions, regulation of cell contraction/migration and cell growth/differentiation. GPCRs interact with G proteins (heterotrimeric GTPases) to synthesize intracellular second messengers, such as diacylglycerol, cyclic AMP, inositol phosphates and calcium ions. Their diverse biological functions range from vision and olfaction to neuronal and endocrine signaling and are involved in many pathological conditions. GRIN2 (G protein-regulated inducer of neurite outgrowth 2), also known as GPRIN2, is a 458 amino acid protein that is expressed in cerebellum and is thought to play a role in neurite outgrowth. GRIN2 interacts with activated G?oand G?, and is encoded by a gene that maps to human chromosome 10q11.22.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GPRIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Documents

Specifications
  • Size:
    100 μg
  • Clonality:
    Polyclonal
  • Purification:
    Purified by Protein A.
  • Antibody type:
    Primary
  • Form:
    liquid
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Antigen name:
    G Protein Regulated Inducer Of Neurite Outgrowth 2
  • Cat. No.:
    BOSSBS-8276R
  • Reactivity:
    Human|Rat|Mouse
  • Gene ID:
    9721
  • Antigen synonyms:
    GRIN2|KIAA0514
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    GPRIN2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.