Order Entry
Denmark
ContactUsLinkComponent
Anti-DHPS1 Rabbit Polyclonal Antibody
Anti-DHPS1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8260R
Supplier:  Bioss
Anti-DHPS1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-8260R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dehydrogenase/reductase (SDR family) member 1
  • Antigen symbol:
    DHPS1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    115817
  • Antigen synonyms:
    Dehydrogenase/reductase SDR family member 1|dehydrogenase/reductase SDR family member 1|DHRS1_HUMAN.|FLJ25430|MGC20204
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

DHRS1 (dehydrogenase/reductase (SDR family) member 1), also known as SDR19C1, is a 313 amino acid protein that belongs to the short-chain dehydrogenases/reductases (SDR) family and likely functions as an oxidoreductase. Abundantly expressed in heart and liver, DHRS1 contains an SDR motif and is encoded by a gene that maps to human chromosome 14q12. Human chromosome 14 houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder ?-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DHPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat