- Antibody type:Primary
- Antigen name:EPH Receptor B1
- Antigen symbol:EPHB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:2047
- Antigen synonyms:Ephrin type-B receptor 2|EphrinB1|Ephrin type-B receptor 3|HEK2|Hek5|NET|Ephrin B3|CAPB|ERK|EPHT3|EphrinB3|PCBC|ELK|Ephrin type-B receptor 1|Ephrin B2|TYRO6|HEK-6|EphrinB2|Tyrosine-protein kinase receptor EPH-2|Tyro5|Ephrin B1|ETK2|MGC87492|EPHB2|EPHB3|DRT
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
EphB1, previously known as Elk (eph like kinase), is a receptor tyrosine kinase of the highly tissue restricted family of eph proteins. EphB1 and other ephB family members are type 1 membrane spanning proteins, comprised of immunoglobulin, fibronectin type III, and cysteine rich subdomains in the ecto domain, and the single uninterrupted cytoplasmic tyrosine kinase domain upstream of a carboxyterminal sterile alpha motif (SAM) domain. EphB family proteins bind ephrins of the B class. EphB1 is expressed predominately in developing neural structures in embryos, and in vascular epithelium of kidney, and other tissues. Upon binding to alternatively oligomerized ephrin B1, EphB1 signals regulation of cell attachment and cell to cell assembly. Members of this protein family are implicated in neuronal and vascular cell targeting.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: EPHB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat