Order Entry
Denmark
ContactUsLinkComponent
Anti-GALT Rabbit Polyclonal Antibody
Anti-GALT Rabbit Polyclonal Antibody
  BOSSBS-3984R
 :  Bioss
Anti-GALT Rabbit Polyclonal Antibody
  BOSSBS-3984R
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Galactose-1-Phosphate Uridylyltransferase
  • Antigen symbol:
    GALT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Gene ID:
    2592
  • Antigen synonyms:
    UDP glucose hexose 1 phosphate uridylyltransferase|GALT_HUMAN.|Galactose 1 phosphate uridyl transferase|Gal 1 P uridylyltransferase|Galactose 1 phosphate uridylyltransferase
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

GALT (Galactose 1 phosphate uridyl transferase) catalyzes the second step of the Leloir pathway of galactose metabolism, namely the conversion of UDP glucose + galactose 1 phosphate to glucose 1 phosphate + UDP galactose. The absence of this enzyme results in classic galactosemia in humans and can be fatal in the newborn period if lactose is not removed from the diet. The pathophysiology of galactosemia has not been clearly defined.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GALT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat