Order Entry
Denmark
ContactUsLinkComponent
Anti-C4 binding protein Rabbit Polyclonal Antibody
Anti-C4 binding protein Rabbit Polyclonal Antibody
Catalog # BOSSBS-15184R
Supplier:  Bioss
Anti-C4 binding protein Rabbit Polyclonal Antibody
Catalog # BOSSBS-15184R
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C4 binding Protein
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    OTTHUMP00000034375|PRP.|C4b-binding protein beta chain|OTTHUMP00000034288|OTTHUMP00000034289|OTTHUMP0000003474|Complement component 4-binding protein beta|Complement component 4 binding protein beta chain|C4BP|Complement component 4 binding protein beta|C4b binding protein beta chain|C4BPB_HUMAN|Proline rich protein|C4BPB
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

C4BPB is a member of a superfamily of proteins composed predominantly of tandemly arrayed short consensus repeats of approximately 60 amino acids. A single, unique beta-chain encoded by this gene assembles with seven identical alpha-chains into the predominant isoform of C4b-binding protein, a multimeric protein that controls activation of the complement cascade through the classical pathway. C4b-binding protein has a regulatory role in the coagulation system also, mediated through the beta-chain binding of protein S, a vitamin K-dependent protein that serves as a cofactor of activated protein C. The genes encoding both alpha and beta chains are located adjacent to each other on human chromosome 1 in the regulator of complement activation gene cluster. Alternative splicing gives rise to multiple transcript variants

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: C4 binding protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat