Order Entry
Denmark
Orders LinkContactUsLinkComponent
NMR sample tube washer
NMR sample tube washer
Supplier:   witeg Labortechnik
NMR sample tube washer
Supplier:  witeg Labortechnik
Certificates
NMR sample tube washer
NMR sample tube washer
2 Options Available Below
Product Details & Documents
Complete with 2 PTFE-adapters to fir 5 cuvets or NMR tubes of 5 and 10 mm diameter in different lengths. Vacuum operated washers allowing the individual washing of 1 to 5 tube, complete with caps and FEPcappilaries (box 1 pce.).
Product Family Options
Product Information
  • Availability
    Price
  • WITG2.900.029
    NMR sample tube washer, acc. to Swiegot, complete, to fit bottles with socket ST 29/32
    NMR sample tube washer, acc. to Swiegot, complete, to fit bottles with socket ST 29/32
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.640,00 kr
    2.640,00 kr
    1 * 1 Items
       2.640,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    WITG2.900.029
  • WITG2.900.032
    NMR sample tube washer, acc. to Swiegot, complete, to fit laboratory bottles with GL 45 thread
    NMR sample tube washer, acc. to Swiegot, complete, to fit laboratory bottles with GL 45 thread
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.640,00 kr
    2.640,00 kr
    1 * 1 Items
       2.640,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    WITG2.900.032