Anti-S100A8 Rabbit Polyclonal Antibody
Catalog # BSBTPB9742
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A8
- Antigen symbol:S100A8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:20201
- Antigen synonyms:p8|CP-10|MRP8|Caga|B8Ag|AI323541|60B8Ag|CFAg
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Protein S100-A8(S100A8) detection. Tested with WB, IHC-P in Mouse;Rat.
Type: Primary
Antigen: S100A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat