Anti-LOX Rabbit Polyclonal Antibody
Catalog # BSBTPB9718
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:lysyl oxidase
- Antigen symbol:LOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4015
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Protein-lysine 6-oxidase(LOX) detection. Tested with WB, ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat