Order Entry
Denmark
ContactUsLinkComponent
 
VOA calibration standards and kits
 :  Restek
 
undefined
VOA calibration standards and kits
 :  Restek

 

  • CAS No.:
    460-00-4
  • Formula:
    C₆H₄BrF
  • Description:
    1-Bromo-4-fluorobenzene (BFB) mix, wit

 

Frequently Bought Together