Capillary columns, Rtx®-5Sil MS (fused silica)
: Restek
Rtx®-5Sil MS columns are optimised for use with conventional MS detectors but for the fastest analyses, they are equally compatible with time-of-flight mass spectrometers.
Crossbond®, selectivity similar to 5% diphenyl/95% dimethyl polysiloxane.
Temperature range: −60 °C to 330/350 °C
- Catalog No:
- REST12723
- REST12724
- REST12753
- REST12754
- REST12726
- REST42703
- REST12739
- REST12740
- REST12721
- REST12794
- REST42702
- REST12705
- REST42704
- REST12736
- REST12737
- REST12738
- Temperature limit:
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+325/350 °C
- −60...+325/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+320/340 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+330/350 °C
- −60...+320/340 °C
- −60...+330/350 °C
Frequently Bought Together