Order Entry
Denmark
ContactUsLinkComponent
Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 65-4714-U100
Supplier:  Tonbo Biosciences
undefined
Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 65-4714-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    No
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

PerCP-Cyanine5.5 Mouse IgG1 Isotype Control (MOPC-21)

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PerCP-Cy5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: