Capillary columns, Rtx®-CLPesticides (fused silica)
Supplier: Restek
Certificates
About this item
Application specific columns for organochlorine pesticides and herbicides. Low bleed ideal for GC-ECD or GC-MS analyses.
Proprietary Crossbond® phases
Temperature range: Stable to 340 °C
Analyse EPA method 8081B, 8082A, 8151A, 504.1, 515, 508.1 and 552.2 compounds without time-consuming column change.
Specifications
- Catalog No:
- REST43301
- REST43101
- REST11120
- REST42102
- REST11123
- REST11321
- REST42302
- REST11136
- REST42301
- REST11337
- REST11139
- REST11140
- REST11340
- REST11323
- REST11324
- REST11126
- REST11326
- REST11123-600
- REST11139-600
- Temperature limit:
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/330 °C
- −60...+320/340 °C
- −60...+320/330 °C
- −60...+300/320 °C
- −60...+320/340 °C
- −60...+300/320 °C
- −60...+300/320 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C
- −60...+320/340 °C