0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pipettes
Micropipettes
Accessories for pipetting system, Microlab 300
Accessories for pipetting system, Microlab 300
HAMI67098-01
:
HAMILTON BONADUZ
Accessories for pipetting system, Microlab 300
HAMI67098-01
:
HAMILTON BONADUZ
Description:
Microlab 300 disposable tip hand probe, tubing kit, incl. 5 ferrules, 5 stainless steel rings, 5 fittings, 5 strain reliefs, 12 gauge tubing and a probe tip
Seq:
02
Pk:
1