Order Entry
Denmark
ContactUsLinkComponent
Anti-CD3, CD4, CD8a, CD45 Mouse Monoclonal Antibody (FITC, violetFluor® 450, PE, PerCP-Cy5.5®) [clone: UCHT1, RPA-T4, RPA-T8, 2D1]
Anti-CD3, CD4, CD8a, CD45 Mouse Monoclonal Antibody (FITC, violetFluor® 450, PE, PerCP-Cy5.5®) [clone: UCHT1, RPA-T4, RPA-T8, 2D1]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD3, CD4, CD8a, CD45 Mouse Monoclonal Antibody (FITC, violetFluor® 450, PE, PerCP-Cy5.5®) [clone: UCHT1, RPA-T4, RPA-T8, 2D1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 91-0101-T100
  • 91-0101-T025
  • Antigen symbol:
  • CD3
  • CD3
  • Antigen name:
  • CD3 Molecule
  • CD3 Molecule
  • Antigen synonyms:
  • CD45|CD3|CD8a|CD4
  • CD45|CD3|CD8a|CD4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • UCHT1, RPA-T4, RPA-T8, 2D1
  • UCHT1, RPA-T4, RPA-T8, 2D1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together