Order Entry
Denmark
ContactUsLinkComponent
Anti-ATP6AP1 Antibody Pair
Anti-ATP6AP1 Antibody Pair
Catalog # ABNOH00000537-AP51
Supplier:  Abnova
Anti-ATP6AP1 Antibody Pair
Catalog # ABNOH00000537-AP51
Supplier:  Abnova

Specifications

  • Antigen name:
    ATPase, H+ transporting, lysosomal accessory protein 1
  • Antigen symbol:
    ATP6AP1
  • Host:
    Multiple
  • Reactivity:
    Human
  • Gene ID:
    537
  • Antigen synonyms:
    Ac45|ATP6IP1|VATPS1|XAP-3|XAP3|16A|ATP6S1|CF2
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

This Set comes with matched Antibody Pair to detect and quantify protein level of human ATP6AP1.

Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-ATP6AP1, IgG1 kappa (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-ATP6AP1 (50 µg);

Type:
Antigen: ATP6AP1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human