Anti-MAP2K3 Mouse Monoclonal Antibody
ABNOH00005606-PW1
: Abnova
- Antigen name:Mitogen-activated protein kinase Kinase 3
- Antigen symbol:MAP2K3
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:5606
- Antigen synonyms:dual specificity mitogen-activated protein kinase kinase 3|SAPKK-2|MEK 3|MAP kinase kinase 3|MAPKK3|SAPK kinase 2|SAPKK2|PRKMK3|stress-activated protein kinase kinase 2|MKK3|MAPKK 3|MAPK/ERK kinase 3
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
- Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-MAP2K3 (300 μg); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-MAP2K3 (50 μg)
Type:
Antigen: MAP2K3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human