Anti-TP53 + TP53 Antibody Pair
ABNODP0014
: Abnova
- Antigen name:Tumor protein p53
- Antigen symbol:P53
- Host:Multiple
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Gene ID:7157
- Antigen synonyms:P53|BCC7|TRP53|LFS1
- Modification:phospho-Ser9
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Protein Phosphorylation Antibody Pair set comes with two Antibodies, one against the TP53 protein, and the other against the specific S9 phosphorylated site of TP53 for use in Proximity Ligation Assay
Antibody pair set content: 1. Phospho-TP53 S9 Rabbit Polyclonal Antibody (20 µl) With 0.09% sodium azide; 2. TP53 Mouse Monoclonal Antibody (40 µg) In 1x PBS, pH 7.2; *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Type:
Antigen: TP53 + TP53
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: