Spray nozzles
Supplier: ENTEGRIS
Jump to:
About this item
PFA spray nozzles with NPT connections for use in applications needing chemical or DI water to be sprayed across a surface.
Molded PFA provides universal chemical compatibility and use in ultrapure and chemically corrosive applications.
5 Options Available Below
Product Details & Documents
PFA spray nozzles with NPT connections for use in applications needing chemical or DI water to be sprayed across a surface.
Molded PFA provides universal chemical compatibility and use in ultrapure and chemically corrosive applications.
Product Family Options
Product Information
- AvailabilityPrice
- ENTE215-66Nozzle for spray gun
- ENTE215-15Nozzle for spray gun
- ENTE215-1Nozzle for spray gun
- ENTE215-28Nozzle for spray gun
- ENTE215-14Nozzle for spray gun