Heat resistant welding gloves, Welder Blue HR
These Gunn-cut gloves feature a palm and back in cowhide split leather and a cuff in split leather lined with canvas. The interior lined with cotton padding.
- US assembly, winged thumb with yoke, fingertips and hand stitching
- Winged thumb and flexible materials used make this glove anatomical, thus providing excellent comfort for use
- Cotton fleece lining increases comfort and heat insulation of the glove
- Split leather treated against the heat for excellent resistance to contact and radiant heat
- Reinforced palm and thumb in split leather increases the glove's durability
- Split leather cuff protects a large part of the forearm against mechanical hazards
Ideal for welding applications and heavy handlings.
: EN 388:2016 (3132X), EN 407 (413X4X); Cat. II
- Catalog No:
- 112-1384
- 112-1379
- Model:
- Welder Blue HR
- Welder Blue HR
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- EN norm:
- EN 388:2016 (3132X), EN 407 (413X4X)
- EN 388:2016 (3132X), EN 407 (413X4X)
Frequently Bought Together