Heat sealing foil, Gas PermASeal™
Non woven gas permeable and peelable seal for PP and PS plates. Suitable for short term incubation, agriculture and seed storage, insect storage, cell culture.
- Pierceable and peelable
- Barriers to solid components
- Porous and opaque
- Seal integrity range: −20 to +80 °C
- Not certified free from nucleases and DNA
Frequently Bought Together