Heat sealing foil, PierceASeal Foil PS™
Leverandør: iST Scientific Ltd
Certifikater
Om denne vare
Pierceble and peelable, high grade foil for polystyrene plates with good solvent resistance including DMSO. Suitable for PCR low temperature compound storage, short term room temperature compound storage.
- Peelable to polystyrene plates only
- Multiple sealing and resealing permissible
- Easy non sealing surface identification
- Seal integrity range: −20 to 110 °C
Frequently Bought Together