Mouse Isotype Control (Unconjugated)
BSENM-1249-100
- Antibody type:Control
- Antigen symbol:IgG
- Clonality:Monoclonal
- Clone:X63
- Conjugation:Unconjugated
- Host:Mouse
- Size:100 µg
- Pk:100 µG
X63 is a control mouse IgG antibody with no known reactivity. The antibody has been used successfully a negative control in westerns, all forms of IHC, IP and FACS
No staining has ever been identified with this immunoglobulin demonstrating its non-specific value as a control.
Application Information:
Recommended for use as a control for Western Blotting, immunohistochemistry and FACS at a concentration equal to that of the test antibody. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Control
Antigen: Unknown
Clonality: Monoclonal
Clone: X63
Conjugation: Unconjugated
Epitope: This control IgG has no known binding ability.
Host: Mouse
Isotype:
Reactivity: