Order Entry
Denmark
ContactUsLinkComponent
Anti-Ubiquitin Rabbit Polyclonal Antibody
Anti-Ubiquitin Rabbit Polyclonal Antibody
Catalog # BSENR-182-250
Supplier:  Biosensis
undefined
Anti-Ubiquitin Rabbit Polyclonal Antibody
Catalog # BSENR-182-250
Supplier:  Biosensis

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin
  • Antigen symbol:
    Ubi-1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Baboon,
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    Multiple points of reactivity likely because of the polyclonal nature of the antibody
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Antigen synonyms:
    RPS27A|UBA52|UBC|UBB
  • Immunogen:
    Ubiquitin isolated from cow erythrocytes and conjugated to chicken gammaglobulins with glutaraldehyde.
  • Size:
    250 µg
  • Pk:
    250 µG

Specifications

About this item

Ubiquitin is a highly conserved 76 amino acid protein with an estimated molecular weight of 8.56 kDa which has a central role in regulated protein degradation. It is a protein modifier which can be covalently attached to target lysines either as a monomer or as a lysine-linked polymer. Several types of polymeric chains can be formed depending on the lysine used for the assembly. Attachment to proteins as a polymer leads to their degradation by the 26S proteosome; a complex, multicatalytic cytosolic and nuclear protease. Attachment to proteins as a monomer or as an alternatively linked polymer does not lead to proteasomal degradation and may be required for numerous functions, including maintenance of chromatic structure, regulation of gene expression, stress response, ribosome biogenesis and DNA repair. Ubiquitin is synthesized as a polyubiquitin precursor with exact head to tail repeats, the number of repeats of which differ between species and strains. In some species there is a final amino-acid after the last repeat, here in bovine a Cys. Some ubiquitin genes contain a single copy of ubiquitin fused to a ribosomal protein (either L40 or S27a).

The specificity of this antibody has been determined by indirect ELISA against ubiquitin conjugated to keyhole limpet haemocyanin. The antibody does not react with kehole limpet haemocyanin alone. Specificity has also been demonstrated by WB against endometrial tissue homogenates.
Application Information:
This antibody is recommended for IHC, WB and immunoprecipitation. This antibody can be used for labelling formalin-fixed, paraffin-embedded tissue sections at a dilution range of 1:150 to 1:300. This antibody is also useful for investigating neurodegenerative diseases such as Alzheimer's disease and Parkinson's disease. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ubiquitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity: Human, Baboon, Rat