Order Entry
Denmark
ContactUsLinkComponent
Electrochemistry kit
Electrochemistry kit
Catalog # BINH1002711
Supplier:  3B Scientific
undefined
Electrochemistry kit
Catalog # BINH1002711
Supplier:  3B Scientific
Specifications
  • Description:
    Equipment set for electrochemistry, 12 pieces
  • Pk:
    1
Specifications
About this item
Equipment set inclusive of digital multimeter for measuring electrochemical potentials of various metals in experiments.
Delivery: Kit supplied with a digital multimeter with 2 cables with crocodile clamps, 1× flat trough, 1× copper plate, 1× zinc plate, 1× iron plate, 2× nickel plate, 1× aluminium plate, 2× electrolyte-carbon plates.